missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-5-Hydroxytryptophan, 99%

Product Code. 10330164 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10330164 25 g Glass Bottle 25g
10336980 1 g Glass Bottle 1g
10143480 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10330164 Supplier Thermo Scientific Chemicals Supplier No. 148290250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C11H12N2O3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H12N2O3
MDL Number MFCD00064341

Specifications

Arsenic (As) 1ppm max.
Color Beige to White or Gray
Heavy Metals 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation -30.9° to -34.9° (20°C, 589nm) (c=1, H2O) on dry substance
Merck Index 15, 4886
Sulfated Ash 0.3% max.
Water 1% max. (K.F.)
Specific Rotation Condition - 32.50 (20.00°C c=1,H2O on dry basis)
Quantity 25 g
Formula Weight 220.23
Percent Purity 98.5 to 101.5% (on dry substance)
Physical Form Powder
Chemical Name or Material L-5-Hydroxytryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.