missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress L-4-Oxalysine hydrochloride
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Molecular Weight (g/mol) | 184.62 g/mol |
| Shipping Condition | Room temperature |
| For Use With (Application) | Neuroscience-Neuromodulation |
| CAS | 118021-35-5 |
| Product Type | Reference compound |
| Purity | 97.1% |
| Target | Fungal |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?