missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-368,899 hydrochloride
GREENER_CHOICE

Product Code. 30001391
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 30001391

Brand: Tocris Bioscience™ 2641/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, non-peptide oxytocin receptor antagonist

Potent, non-peptide and orally active oxytocin receptor antagonist (IC50 = 8.9 nM) that displays > 40-fold selectivity over vasopressin V1a and V2 receptors (IC50 values are 370 and 570 nM respectively). Antagonizes oxytocin-induced uterine contractions in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160312-62-9
Molecular Formula C26H43ClN4O5S2

Specifications

Quantity 1 mg
Formula Weight 591.23
Purity >97%
Molecular Weight (g/mol) 591.223
Chemical Name or Material L-368, 899 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.