missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ L-365,260

Product Code. 18095672
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095672 10 mg 10mg
18065392 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095672 Supplier R&D Systems™ Supplier No. 2767/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective CCK2 antagonist

Selective cholecystokinin receptor 2 (CCK2) antagonist (IC50 values are 2 and 280 nM at CCK2 and CCK1 receptors respectively) that is inactive at a range of other receptors including opiate, muscarinic acetylcholine, α- and β adrenergic, histamine, angiotensin and bradykinin receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118101-09-0
Molecular Formula C24H22N4O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 398.46
Purity >99%
Molecular Weight (g/mol) 398.466
Chemical Name or Material L-365, 260
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.