missing translation for 'onlineSavingsMsg'
Learn More

L-3-Phosphoglyceric Acid 13C3 Sodium Salt, TRC

Product Code. 30009164
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009164 2.5 mg 2.5mg
30006447 1 mg 1mg
30008098 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009164 Supplier Toronto Research Chemicals Supplier No. TRCP3580082.5MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C3H6O7P • x(Na)
IUPAC Name sodium (S)-2-(carboxy-13C)-2-hydroxyethyl-1,2-13C2 hydrogen phosphate
SMILES O=[13C](O)[13C@]([13C](OP(O)([O-])=O)([H])[H])(O)[H].[Na+]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C3H7O7P.Na/c4-2(3(5)6)1-10-11(7,8)9;/h2,4H,1H2,(H,5,6)(H2,7,8,9);/q;+1/p-1/t2-;/m0./s1/i1+1,2+1,3+1;
Percent Purity >95%
Chemical Name or Material L-3-Phosphoglyceric Acid 13C3 Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.