missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-3-Phenyllactic Acid, TRC

Product Code. 30425307
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425307 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425307 Supplier Toronto Research Chemicals Supplier No. TRCP335553500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-(-)-3-Phenyllactic Acid
CAS 20312-36-1
Molecular Formula C9H10O3
Quantity 500 mg
Synonym (S)-a-Hydroxy-benzenepropanoic Acid, (S)-3-Phenyllactic Acid (8CI), (aS)-a-Hydroxybenzenepropanoic Acid, (-)-2-Hydroxy-3- phenylpropanoic Acid, (-)-2-Hydroxy-3-phenylpropionic Acid, (-)-3-Phenyllactic Acid, (-)-b-Phenyllactic Acid, (2S)-2-Hydroxy-3-phenylpropanoic Acid, (S)-2-Hydroxy-3-phenylpropanoic Acid, (S)-2-Hydroxy-3-phenylpropionic Acid, (S)-3-Phenyllactic Acid, (S)-Phenyllactic Acid, (S)-b-Phenyllactic Acid, 3-(S)-Phenyl lactic Acid, L-2-Hydroxy-3-phenylpropionic Acid, L-3-Phenyllactic Acid, L-Phenyllactic Acid, L-b-Phenyllactic Acid, S-(-)-3-Phenyllactic Acid, b-Phenyl-L-lactic Acid
InChI Formula InChI=1S/C9H10O3/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)/t8-/m0/s1
SMILES O[C@H](C(O)=O)CC1=CC=CC=C1
IUPAC Name (2S)-2-hydroxy-3-phenylpropanoic acid
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.