missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-2-Phenylglycine methyl ester hydrochloride, 97%

Product Code. 15437946
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437946 25 g 25g
15447946 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437946 Supplier Thermo Scientific Alfa Aesar Supplier No. H62874.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15028-39-4 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15028-39-4
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00077158
InChI Key DTHMTBUWTGVEFG-QRPNPIFTSA-N
Synonym h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride
PubChem CID 11217910
IUPAC Name methyl (2S)-2-amino-2-phenylacetate;hydrochloride
SMILES COC(=O)C(C1=CC=CC=C1)N.Cl

Specifications

Melting Point 200°C (decomposition)
Quantity 25 g
Formula Weight 201.65
Optical Rotation +118° (c=1 in Water)
Percent Purity 97%
Chemical Name or Material L-(+)-2-Phenylglycine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.