Learn More
L-(+)-2-Aminobutyric acid, 98+%
Description
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-(+)-2-Aminobutyric acid is used in the biosynthesis of nonribosomal peptides. It acts as a receptor antagonist. It is also used as a chiral reagent. Further, it is used in the determination of substrate of glutamyl cysteine acid synthase. In addition to this, it is also utilized as a drug intermediate.
Solubility
Soluble in water and acetic acid. Slightly soluble in alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 1720935 |
| Merck Index | 14,428 |
| Solubility Information | Soluble in water and acetic acid. Slightly soluble in alcohol and ether. |
| Optical Rotation | +21° (c=4.8 in 6N HCl) |
| Formula Weight | 103.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | L-(+)-2-Aminobutyric acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.