missing translation for 'onlineSavingsMsg'
Learn More

L-2-Aminoadipic acid, 98%

Product Code. 11358825
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358825 1 g 1g
11368825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358825 Supplier Thermo Scientific Alfa Aesar Supplier No. L13924.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1118-90-7 | C6H11NO4 | 161.16 g/mol

L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-2-Aminoadipic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in 1N HCl (50 mg/ml) and water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-90-7
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00002636
InChI Key OYIFNHCXNCRBQI-BYPYZUCNSA-N
Synonym l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
PubChem CID 92136
ChEBI CHEBI:37023
IUPAC Name (2S)-2-aminohexanedioic acid
SMILES N[C@@H](CCCC(O)=O)C(O)=O

Specifications

Melting Point ∽206°C (decomposition)
Quantity 1 g
Beilstein 1724348
Solubility Information Soluble in 1N HCl (50mg/ml) and water (slightly).
Optical Rotation +25° (c=2 in 5N HCl)
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material L-2-Aminoadipic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.