missing translation for 'onlineSavingsMsg'
Learn More

L-2,4-Diaminobutyric acid dihydrochloride, 98+%, may cont. up to ca 10% monohydrochloride

Product Code. 11332437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332437 5 g 5g
11322437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332437 Supplier Thermo Scientific Alfa Aesar Supplier No. L10093.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1883-09-6 | C4H12Cl2N2O2 | 191.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1883-09-6
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00064561
InChI Key CKAAWCHIBBNLOJ-QTNFYWBSSA-N
Synonym l-2,4-diaminobutyric acid dihydrochloride, s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diaminobutyric acid dihydrochloride, h-dab-oh.2hcl, 2,4-diaminobutyric acid dihydrochloride, 2s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diamino-n-butyric acid dihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, 2s, d-dab.2hcl, 2s-2,4-diaminobutanoic acid, chloride, chloride
PubChem CID 2724265
IUPAC Name (2S)-2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Melting Point ∼200°C
Quantity 5 g
Beilstein 5763078
Optical Rotation +18° (c=1 in 5N HCl)
Formula Weight 191.06
Percent Purity ≥98%
Assay may cont. up to ≈10% monohydrochloride
Chemical Name or Material L-2,4-Diaminobutyric acid dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.