missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-165041, MedChemExpress
Descrizione
L-165041 is a cell permeable PPARδ agonist, with Kis of 6 nM and appr 730 nM for PPARδ and PPARγ, respectively, and induces adipocyte differentiation in NIH-PPARδ cells.
Specifica
Specifica
| Chemical Name or Material | L-165041 |
| CAS | 79558-09-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H26O7 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (124.24 mM; Need ultrasonic) |
| SMILES | O=C(O)COC(C=C1)=CC=C1OCCCOC2=CC=C(C(C)=O)C(O)=C2CCC |
| Molecular Weight (g/mol) | 402.44 |
| Formula Weight | 402.44 |
| Vedi altri risultati |
Research purposes only
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?