missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-165,041, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent PPARδ agonist (Ki = 6 nM); displays > 100-fold selectivity for both mouse and human PPARδ receptors over other subtypes. Raises plasma cholesterol levels in vivo, in insulin-resistant db/db mice. Neuroprotective in models of cerebral infarction and Parkinson's disease.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 402.44 |
| Percent Purity | >98% |
| Chemical Name or Material | L-165, 041 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction