missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ L-15 (Leibovitz), GlutaminePlus, no phenol red
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |
| Purity or Quality Grade | 0.2 μm filtered |
| Content And Storage | 2 - 8 °C, protected from light |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?