missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kynurenic acid sodium, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Kynurenic acid sodium, an endogenous tryptophan metabolite, is a broad-spectrum antagonist targeting NMDA, glutamate, α7 nicotinic acetylcholine receptor. Kynurenic acid sodium is also an agonist of GPR35/CXCR8.
Specifications
Specifications
| Chemical Name or Material | Kynurenic acid sodium |
| Physical Form | Solid |
| CAS | 2439-02-3 |
| Color | Gray |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C10H6NNaO3 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 50 mg/mL (236.80 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.47 mM; Need ultrasonic) |
| SMILES | O=C(C1=NC2=CC=CC=C2C(O)=C1)O[Na] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction