missing translation for 'onlineSavingsMsg'
Learn More

Kynurenic acid, MedChemExpress

Product Code. 16451526
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16451526

Brand: Medchem Express HY100806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kynurenic acid, an endogenous tryptophan metabolite, is a broad-spectrum antagonist targeting NMDA, glutamate, α7 nicotinic acetylcholine receptor. Kynurenic acid is also an agonist of GPR35/CXCR8.

Kynurenic acid, an endogenous tryptophan metabolite, is a broad-spectrum antagonist targeting NMDA, glutamate, α7 nicotinic acetylcholine receptor. Kynurenic acid is also an agonist of GPR35/CXCR8.

Specifications

Chemical Name or Material Kynurenic acid
CAS 492-27-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H7NO3
Synonym Quinurenic acid
Solubility Information DMSO : 9 mg/mL (47.58 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC2=CC=CC=C2C(O)=C1)O
Molecular Weight (g/mol) 189.17
Quantity 500 mg
Formula Weight 189.17
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.