missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KYNU Monoclonal Antibody (OTI5E4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526643
This item is not returnable.
View return policy
Description
KYNU Monoclonal Antibody for Western Blot, IHC (P)
Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| KYNU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16719 | |
| Kynu | |
| Human recombinant protein fragment corresponding to amino acids 1-216 of KYNU produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5E4 | |
| Unconjugated | |
| Kynu | |
| 4432411A05Rik; HAMAP-Rule:MF_03017}; KYNU; kynureninase; kynureninase (L-kynurenine hydrolase); kynureninase {ECO:0000255; kynureninase-like protein; KYNUU; L-kynurenine hydrolase; L-kynurenine hydrolase {ECO:0000255 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8942 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction