missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KYNU Monoclonal Antibody (KAA446_19F4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA555692
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence of this protein is as follows: DTVQRIAAEL KCHPTDERVA LHLDEEDKLR HFRECFYIPK IQDLPPVDLS LVNKDENAIY FLGNSLGLQP KMVKTYLEEE LDKWAKIAAY GHEVGKRPWI TGDESIVGLM KDIVGANEKE IALMNALTVN LHLLMLSFFK PTPKRYKILL EAKAFPSDHY AIESQLQLHG LNIEESMRMI KPREGEETLR IEDILEVIEK
Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| KYNU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q16719 | |
| Kynu | |
| Recombinant human KYNU (immunogen range is 11-210). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| KAA446_19F4 | |
| Unconjugated | |
| Kynu | |
| 4432411A05Rik; HAMAP-Rule:MF_03017}; KYNU; kynureninase; kynureninase (L-kynurenine hydrolase); kynureninase {ECO:0000255; kynureninase-like protein; KYNUU; L-kynurenine hydrolase; L-kynurenine hydrolase {ECO:0000255 | |
| Mouse | |
| Protein A | |
| RUO | |
| 8942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido