Learn More
Masterflex™ Kynar™ Reducing Connectors, PVDF
Click to view available options
:
For 1/2 x 3/8 in. tubing I.D.
Description
- Designed for quick and easy assembly without tools
- Excellent chemical resistance
- Temp range: -62 to 129°C
- Sterilize by ethylene oxide or autoclave
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PVDF |
| For Use With | 1/2 x 3/8 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Port Size | 1/4 in. orifice; 2-1/2 x 11/16 in. |