missing translation for 'onlineSavingsMsg'
Learn More

KY1220, MedChemExpress

Product Code. 16494766
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494766 100 mg 100mg
16492826 25 mg 25mg
16480077 50 mg 50mg
16473688 10 mg 10mg
16466427 5 mg 5mg
16400076 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16494766 Supplier Medchem Express Supplier No. HY102028
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KY1220 is a compound that destabilizes both β-catenin and Ras, via targeting the Wnt/β-catenin pathway; with an IC50 of 2.1 μM in HEK293 reporter cells.

KY1220 is a compound that destabilizes both β-catenin and Ras, via targeting the Wnt/β-catenin pathway

Specifications

Chemical Name or Material KY1220
CAS 292168-79-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H10N4O3S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (318.15 mM)
SMILES O=C(/C(N1)=C/C2=CC=CN2C3=CC=C([N+]([O-])=O)C=C3)NC1=S
Molecular Weight (g/mol) 314.32
Formula Weight 314.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.