missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies KY02111

Product Code. 17130092
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130092 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130092 Supplier STEMCELL Technologies Supplier No. 72582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WNT pathway inhibitor

  • KY02111 promotes the differentiation of pluripotent stem cells (PSCs) to cardiomyocytes by inhibiting canonical WNT signaling in a manner distinct from that of other WNT inhibitors (Minami et al.)
  • Promotes cardiomyocyte differentiation of human and mouse PSCs in combination with BIO, CHIR99021, and XAV939 (Minami et al.)

Specifications

CAS 1118807-13-8
Quantity 5 mg
Molecular Formula C18H17ClN2O3S
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.