missing translation for 'onlineSavingsMsg'
Learn More

KY02111, MedChemExpress

Product Code. p-200099620
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16432027

Brand: Medchem Express HY13815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KY02111 is a canonical WNT signaling (β-catenin) inhibitor which promotes differentiation of hPSCs to cardiomyocytes. KY02111 can be used for the research of human cardiomyocyte regeneration.

KY02111 is a canonical WNT signaling (β-catenin) inhibitor which promotes differentiation of hPSCs to cardiomyocytes. KY02111 can be used for the research of human cardiomyocyte regeneration.

Specifications

Chemical Name or Material KY02111
CAS 1118807-13-8
Color White
Purity Grade Notes Research
Molecular Formula C18H17ClN2O3S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (88.44 mM; Need ultrasonic)
SMILES O=C(NC1=NC2=CC=C(Cl)C=C2S1)CCC3=CC=C(OC)C(OC)=C3
Molecular Weight (g/mol) 376.86
Formula Weight 376.86
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.