missing translation for 'onlineSavingsMsg'
Learn More

KY02111, MedChemExpress

Product Code. 16432027
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432027 50 mg 50mg
16425878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432027 Supplier Medchem Express Supplier No. HY13815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KY02111 is a canonical WNT signaling (β-catenin) inhibitor which promotes differentiation of hPSCs to cardiomyocytes. KY02111 can be used for the research of human cardiomyocyte regeneration.

KY02111 is a canonical WNT signaling (β-catenin) inhibitor which promotes differentiation of hPSCs to cardiomyocytes. KY02111 can be used for the research of human cardiomyocyte regeneration.

Specifications

Chemical Name or Material KY02111
CAS 1118807-13-8
Color White
Purity Grade Notes Research
Molecular Formula C18H17ClN2O3S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (88.44 mM; Need ultrasonic)
SMILES O=C(NC1=NC2=CC=C(Cl)C=C2S1)CCC3=CC=C(OC)C(OC)=C3
Molecular Weight (g/mol) 376.86
Formula Weight 376.86
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.