missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KY 02111
Description
KY 02111 promotes differentiation of human ESCs and iPSCs into cardiomyocytes when used after day three in culture; differentiation efficiency increases when used after initial culture with CHIR 99021 (Cat. No. 4423). Induces downregulation of Wnt signaling target genes; inhibits canonical Wnt signaling in a manner distinct from other known Wnt inhibitors.
Specifications
Specifications
| Chemical Name or Material | N-(6-Chloro-2-benzothiazolyl)-3,4-dimethoxybenzenepropanamide |
| CAS | 1118807-13-8 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C18H17ClN2O3S |
| Formula Weight | Observed MW: 376.86 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?