missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KXD1 Monoclonal Antibody (4C9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522554
This item is not returnable.
View return policy
Description
Peptide Sequence: FRRIRTLKGK LARQHPEAFS HIPEASFLEE EDEDPIPPST TTTIATSEQS TGSCDTSPDT VSPSLSPGFE DLSHVQAGSP AINGRSQTDD EEMTGE.
The function of the C19orf50 protein has not yet been determined.
Specifications
| KXD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BQD3 | |
| KXD1 | |
| KXD1 (NP_076974, 81 a.a. ~ 176 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4C9 | |
| Unconjugated | |
| KXD1 | |
| BORCS4; C19orf50; KXD1; KXDL; KxDL motif containing 1; kxDL motif-containing protein 1; MST096; MSTP096; UPF0459 protein C19orf50 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79036 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction