missing translation for 'onlineSavingsMsg'
Learn More

KW-8232 free base, MedChemExpress

Product Code. 16470609
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470609 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16470609 Supplier Medchem Express Supplier No. HY100304
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress KW-8232 free base, an orally active anti-osteoporotic agent, and can reduces the biosynthesis of PGE2.

KW-8232 free base, an orally active anti-osteoporotic agent, and can reduces the biosynthesis of PGE2.

Specifications

Chemical Name or Material KW-8232 free base
CAS 170365-25-0
Color Pink
Purity Grade Notes Research
Molecular Formula C36H37ClN4O3
Quantity 1 mg
SMILES O=C(C(N1CCN(C)C)=C(C(C2=CC=C(O)C=C2)C3=CC=C(O)C=C3)C4=C1C=CC=C4)N5CCN(C6=CC=CC=C6Cl)CC5
Molecular Weight (g/mol) 609.16
Formula Weight 609.16
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.