missing translation for 'onlineSavingsMsg'
Learn More

KW-2449, MedChemExpress

Product Code. 16443838
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443838 10 mg 10mg
16490207 50 mg 50mg
16454876 100 mg 100mg
16406567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443838 Supplier Medchem Express Supplier No. HY10339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

Especificaciones

Chemical Name or Material KW-2449
Physical Form Powder
CAS 1000669-72-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (150.42 mM)
SMILES O=C(C1=CC=C(/C=C/C2=NNC3=C2C=CC=C3)C=C1)N4CCNCC4
Molecular Weight (g/mol) 332.4
Formula Weight 332.4
Percent Purity 99.72%
Grade Research
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.