missing translation for 'onlineSavingsMsg'
Learn More

KW-2449, MedChemExpress

Product Code. 16490207
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490207 50 mg 50mg
16454876 100 mg 100mg
16443838 10 mg 10mg
16406567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490207 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY10339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

KW-2449 is a multi-targeted kinase inhibitor of FLT3, ABL, ABLT315I and Aurora kinase with IC50s of 6.6, 14, 4 and 48 nM, respectively.

Specifications

Chemical Name or Material KW-2449
Physical Form Powder
CAS 1000669-72-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (150.42 mM)
SMILES O=C(C1=CC=C(/C=C/C2=NNC3=C2C=CC=C3)C=C1)N4CCNCC4
Molecular Weight (g/mol) 332.4
Formula Weight 332.4
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.