missing translation for 'onlineSavingsMsg'
Learn More

KvBeta3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18370067 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370067 100 μL 100µL
18305826 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370067 Supplier Novus Biologicals Supplier No. NBP303643100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KvBeta3 Polyclonal specifically detects KvBeta3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KvBeta3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias AKR6A9, K(+) channel subunit beta-3, KCNA3.1B, KCNA3BK+ channel beta-3 subunit, Kv-beta-3, MGC116886, potassium channel, voltage-dependent, beta-3 subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 3, voltage-gated potassium channel subunit beta-3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 285-404 of human KvBeta3 (NP_004723.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9196
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.