missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KvBeta2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80271
This item is not returnable.
View return policy
Description
KvBeta2 Polyclonal specifically detects KvBeta2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| KvBeta2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HKvbeta2, HKvbeta2.1, HKvbeta2.2, K(+) channel subunit beta-2, K+ channel beta-2 subunit, KCNA2BAKR6A5, KCNK2, KV-BETA-2, MGC117289, potassium channel shaker chain beta 2, potassium voltage-gated channel, shaker-related subfamily, beta member 2, voltage-gated potassium channel subunit beta-2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 100%; Bovine: 100%; Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_003627 | |
| KCNAB2 | |
| Synthetic peptide directed towards the middle region of human KCNAB2. Peptide sequence: WGGKAETERGLSRKHIIEGLKASLERLQLEYVDVVFANRPDPNTPMEETV | |
| 100 μL | |
| Signal Transduction | |
| 8514 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction