missing translation for 'onlineSavingsMsg'
Learn More

Kv9.3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05922-100ul

 View more versions of this product

Product Code. 18648385

  • £255.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Kv9.3 Polyclonal antibody specifically detects Kv9.3 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kv9.3
Polyclonal
Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate
delayed-rectifier K(+) channel alpha subunit 3, KV9.3, MGC9481, potassium voltage-gated channel subfamily S member 3, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3, Shab-related delayed-rectifier K+ channel alpha subunit 3, voltage-gated potassium channel protein Kv9.3, voltage-gated potassium channel subunit Kv9.3
Produced in rabbits immunized with E. coli-derived Human Kv9.3 fragment.
100 μg
Neuroscience
3790
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.