missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kv8.2 Polyclonal antibody specifically detects Kv8.2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Kv8.2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | KV11.1, Kv8.2, MGC120515, potassium channel, subfamily V, member 2, potassium voltage-gated channel subfamily V member 2, RCD3B, voltage-gated potassium channel Kv8.2, Voltage-gated potassium channel subunit Kv8.2 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human Kv8.2. The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?