missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv7.4 Antibody (S43/6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12899-0.025mg
This item is not returnable.
View return policy
Description
Kv7.4 Monoclonal specifically detects Kv7.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Microarray.
Specifications
| Kv7.4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| DFNA2, DFNA2A, KQT-like 4, KV7.4, potassium channel KQT-like 4, Potassium channel subunit alpha KvLQT4, potassium voltage-gated channel subfamily KQT member 4, potassium voltage-gated channel, KQT-like subfamily, member 4, Voltage-gated potassium channel subunit Kv7.4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9132 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| S43/6 | |
| Unconjugated | |
| NP_004691.2 | |
| KCNQ4 | |
| Fusion protein amino acids 2-77 of human KCNQ4 | |
| 0.025 mg | |
| Primary | |
| Detects approx 77kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction