missing translation for 'onlineSavingsMsg'
Learn More

Kv7.4 Antibody (S43/6), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18418312 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418312 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418312 Supplier Novus Biologicals Supplier No. NBP212899JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kv7.4 Monoclonal antibody specifically detects Kv7.4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv7.4
Applications Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray
Classification Monoclonal
Clone S43/6
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias DFNA2, DFNA2A, KQT-like 4, KV7.4, potassium channel KQT-like 4, Potassium channel subunit alpha KvLQT4, potassium voltage-gated channel subfamily KQT member 4, potassium voltage-gated channel, KQT-like subfamily, member 4, Voltage-gated potassium channel subunit Kv7.4
Host Species Mouse
Immunogen Fusion protein amino acids 2-77 of human KCNQ4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience, Potassium Channels
Primary or Secondary Primary
Gene ID (Entrez) 9132
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.