missing translation for 'onlineSavingsMsg'
Learn More

Kv7.2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18353474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353474 20 μL 20µL
18386096 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18353474 Supplier Novus Biologicals Supplier No. NBP30361620UL

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Kv7.2 Polyclonal specifically detects Kv7.2 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Kv7.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BFNC, BFNS1, EBN, EBN1, EIEE7, ENB1, HNSPC, KCNA11, KQT-like 2, Kv7.2, KVEBN1, neuroblastoma-specific potassium channel protein, Neuroblastoma-specific potassium channel subunit alpha KvLQT2, potassium voltage-gated channel subfamily KQT member 2, potassium voltage-gated channel, KQT-like subfamily, member 2, Voltage-gated potassium channel subunit Kv7.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 254-393 of human Kv7.2 (NP_742107.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3785
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.