missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv7.1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | Kv7.1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18345235
|
Novus Biologicals
NBP3-03610-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324936
|
Novus Biologicals
NBP3-03610-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv7.1 Polyclonal specifically detects Kv7.1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Kv7.1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3784 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| ATFB3, IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1, JLNS1, KCNA8FLJ26167, KCNA9Kv1.9, kidney and cardiac voltage dependend K+ channel, KQT-like 1, Kv7.1, KVLQT1ATFB1, LQT, LQT1, potassium voltage-gated channel subfamily KQT member 1, potassium voltage-gated channel, KQT-like subfamily, member 1, RWS, slow delayed rectifier channel subunit, SQT2, Voltage-gated potassium channel subunit Kv7.1, WRS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-549 of human Kv7.1 (NP_861463.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title