missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv7.1 Antibody (5E12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37538-0.025ml
This item is not returnable.
View return policy
Description
Kv7.1 Monoclonal antibody specifically detects Kv7.1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Kv7.1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 5E12 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| ATFB3, IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1, JLNS1, KCNA8FLJ26167, KCNA9Kv1.9, kidney and cardiac voltage dependend K+ channel, KQT-like 1, Kv7.1, KVLQT1ATFB1, LQT, LQT1, potassium voltage-gated channel subfamily KQT member 1, potassium voltage-gated channel, KQT-like subfamily, member 1, RWS, slow delayed rectifier channel subunit, SQT2, Voltage-gated potassium channel subunit Kv7.1, WRS | |
| Purified recombinant fragment of human Kv7.1 expressed in E. Coli. | |
| 0.025 mL | |
| Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 3784 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction