missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv6.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17027
This item is not returnable.
View return policy
Description
Kv6.1 Polyclonal specifically detects Kv6.1 in Human samples. It is validated for Western Blot.
Specifications
| Kv6.1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| KCNG, KH2, kH2K13, KV6.1, MGC12878, potassium channel KH2, potassium channel Kv6.1, potassium voltage-gated channel subfamily G member 1, potassium voltage-gated channel, subfamily G, member 1, Voltage-gated potassium channel subunit Kv6.1 | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Porcine (94%), Rat (91%), Bovine (94%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.87mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9UIX4 | |
| KCNG1 | |
| Recombinant protein encompassing a sequence within the center region of human KCNG1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 3755 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction