missing translation for 'onlineSavingsMsg'
Learn More

Kv5.1 Antibody (1B2-7B6), Novus Biologicals™

Product Code. 18349278 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349278 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349278 Supplier Novus Biologicals Supplier No. H00003754M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kv5.1 Monoclonal antibody specifically detects Kv5.1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv5.1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B2-7B6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH26110
Gene Alias IK8, kH1, KV5.1, MGC33316, potassium channel KH1, potassium channel Kv5.1, potassium voltage-gated channel subfamily F member 1, potassium voltage-gated channel, subfamily F, member 1, voltage-gated potassium channel subunit Kv5.1
Host Species Mouse
Immunogen KCNF1 (AAH26110, 1 a.a. ~ 494 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDGSGERSLPEPGSQSSAASDDIEIVVNVGGVRQVLYGDLLSQYPETRLAELINCLAGGYDTIFSLCDDYDPGKREFYFDRDPDAFKCVIEVYYFGEVHMKKGICPICFKNEMDFWKVDLKFLDDCCKSHLSEKREELEEIARRVQLILDDLGVDAAEGRWRRCQKCVWKFLEKPESSCPARVVAVLSFLLILVSSVVMCMGTIPELQVLDAEGNRVEHPTLENVETACIGWFTLEYLLRLFSSPNKLHFALSFMNIVDVLAILPFYVSLTLTHLGARMMELTNVQQAVQALRIMRIARIFKLARHSSGLQTLTYALKRSFKELGLLLMYLAVGIFVFSALGYTMEQSHPETLFKSIPQSFWWAIITMTTVGYGDIYPKTTLGKLNAAISFLCGVIAIALPIHPIINNFVRYYNKQRVLETAAKHELELMELNSSSGGEGKTGGSRSDLDNLPPEPAGKEAPSCSSRLKLSHSDTFIPLLTEEKHHRTRLQSCK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3754
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.