missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv4.2 Antibody (5B11B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £435.00
Specifications
| Antigen | Kv4.2 |
|---|---|
| Clone | 5B11B9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18117699
|
Novus Biologicals
NBP2-52497 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679137
|
Novus Biologicals
NBP2-52497-0.025mg |
0.025 mg |
£163.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv4.2 Monoclonal specifically detects Kv4.2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Kv4.2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS with 0.05% Sodium Azide | |
| 3751 | |
| Purified recombinant fragment of human KCND2 (AA: 27-184) expressed in E. Coli. | |
| Primary | |
| 70.5 kDa |
| 5B11B9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
| KCND2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title