missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv3.4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | Kv3.4 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18448561
|
Novus Biologicals
NBP1-89887 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18457681
|
Novus Biologicals
NBP1-89887-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv3.4 Polyclonal antibody specifically detects Kv3.4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Kv3.4 | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol | |
| 3749 | |
| IgG | |
| Immunogen affinity purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ISSVCVSSYRGRKSGNKPPSKTCLKEEMAKGEASEKIIINVGGTRHETYRSTLRTLPGTRLAWLADPD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title