missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV3.3 (KCNC3) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577619
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes.
Specifications
| KV3.3 (KCNC3) | |
| Polyclonal | |
| Unconjugated | |
| KCNC3 | |
| KCNC3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117101, 16504, 3748 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q01956, Q14003, Q63959 | |
| Kcnc3 | |
| Peptide KSPITPGSRGRYSRDRAC, corresponding to amino acid residues 701-718 of rat Kv 3.3 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction