Learn More
Kv3.3 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05792APC
This item is not returnable.
View return policy
Description
Kv3.3 Polyclonal antibody specifically detects Kv3.3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kv3.3 | |
| APC | |
| PBS |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Purified |