missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV3.2 (KCNC2) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599182
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNC2.
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels. The subunits vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits, which include the KV1, KV2, KV3, KV4 and KV9 proteins, as well as accessory subunits that modify the gating properties of the coexpressed KV subunits. KV3.2 is a multipass membrane protein that regulates the voltage-dependent K+ permeability of excitable membranes. The tail of KV3.2 may be influential in the targeting of the channel to specific subcellular compartments and/or the regulation of channel activity.
Specifications
| KV3.2 (KCNC2) | |
| Polyclonal | |
| Unconjugated | |
| KCNC2 | |
| AW047325; B230117I07; KCNC2; KShIIIA; Kv3.2; Potassium channel voltage-gated Shaw-related subfamily C member 2; potassium channel, voltage gated Shaw related subfamily C, member 2; potassium channel, voltage gated Shaw-related subfamily C, member 2; potassium voltage gated channel, Shaw-related subfamily, member 2; potassium voltage-gated channel subfamily C member 2; potassium voltage-gated channel, Shaw-related subfamily, member 2; shaw-like potassium channel; Voltage-gated potassium channel Kv3.2; voltage-gated potassium channel subunit Kv3.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246153, 268345, 3747 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22462, Q14B80, Q96PR1 | |
| KCNC2 | |
| A synthesized peptide derived from human KCNC2(Accession Q96PR1), corresponding to amino acid residues G606-L638. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction