missing translation for 'onlineSavingsMsg'
Learn More

KV3.2 (KCNC2) Polyclonal Antibody, Invitrogen™

Product Code. 15944085
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15944085 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15944085 Supplier Invitrogen Supplier No. PA577579

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.

Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. The KV gene family encodes more than 30 proteins that comprise the subunits of the K+ channels. The subunits vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha subunits, which include the KV1, KV2, KV3, KV4 and KV9 proteins, as well as accessory subunits that modify the gating properties of the coexpressed KV subunits. KV3.2 is a multipass membrane protein that regulates the voltage-dependent K+ permeability of excitable membranes. The tail of KV3.2 may be influential in the targeting of the channel to specific subcellular compartments and/or the regulation of channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV3.2 (KCNC2)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 5% sucrose and 0.05% sodium azide; pH 7.4
Gene KCNC2
Gene Accession No. P22462, Q14B80, Q96PR1
Gene Alias Kcnc2
Gene Symbols Kcnc2
Host Species Rabbit
Immunogen Peptide DLGGKRLGIEDAAGLGGPDGK(C), corresponding to amino acid residues 184-204 of rat KV 3.2
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246153, 268345, 3747
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.