missing translation for 'onlineSavingsMsg'
Learn More

Kv3.2 Antibody (S410-17), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18407054 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407054 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407054 Supplier Novus Biologicals Supplier No. NBP259328JF549

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kv3.2 Monoclonal antibody specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv3.2
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone S410-17
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2
Host Species Mouse
Immunogen Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3747
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.