missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv3.2 Antibody (S410-17), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59328AF488
This item is not returnable.
View return policy
Description
Kv3.2 Monoclonal specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Kv3.2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KCNC2 | |
| Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a | |
| 100 μL | |
| Primary | |
| Detects 80-100kDa. Varies due to post-translational modifications. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S410-17 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2 | |
| Mouse | |
| Protein G purified | |
| Signal Transduction | |
| 3747 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction