missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV2.2 (KCNB2) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566815
This item is not returnable.
View return policy
Description
Immunogen sequence: QHISTILLEET PSQGDRPLLG TEVSAPCQGP SKGLSPRFPK QKLFPFSSRE RRSFTEIDTG DDEDFLELP Highest antigen sequence identity to the following orthologs - mouse 79%, rat 81%.
Kcnb2 (Kv2.2) encodes a Voltage-gated K+ channel which are important determinants of neuronal membrane excitability. Moreover, differences in K+ channel expression patterns and densities contribute to the variation in action potential waveforms and repetitive firing patterns evident in different neuronal cell types. The delayed rectifier-type (IK) channels (Kv1.5, Kv2.1, and Kv2.2) are expressed on all neuronal somata and proximal dendrites and are also found in a wide variety on non-neuronal cell types including pancreatic islet, alveolar cells and cardiac myocytes. Kv2.1 and Kcnb2 form distinct populations of K+ channels and these subunits are thought to be primarily responsible for IK in superior cervical ganglion cells. Functionally, Kcnb2 mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient. Diseases associated with KCNB2 include Brugada Syndrome.
Specifications
| KV2.2 (KCNB2) | |
| Polyclonal | |
| Unconjugated | |
| KCNB2 | |
| 9630047L19Rik; BB130875; CDRK; delayed rectifier potassium channel protein; kcnb2; kcnb2.L; kcnb2-a; kcnb2-b; Kv2.2; potassium channel Kv2.2; potassium channel, voltage gated Shab related subfamily B, member 2; potassium channel, voltage gated Shab related subfamily B, member 2 L homeolog; potassium channel, voltage gated Shab-related subfamily B, member 2; potassium voltage gated channel, Shab-related subfamily, member 2; potassium voltage-gated channel subfamily B member 2; potassium voltage-gated channel, Shab-related subfamily, member 2; voltage-gated potassium channel subunit Kv2.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92953 | |
| KCNB2 | |
| Recombinant Human KCNB2. Recombinant protein control fragment (Product #RP-107471). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction