missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV2.1 (KCNB1) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA112658
This item is not returnable.
View return policy
Description
KV2.1 (KCNB1) Polyclonal Antibody for Western Blot
Mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient.
Specifications
| KV2.1 (KCNB1) | |
| Polyclonal | |
| Unconjugated | |
| Kcnb1 | |
| Delayed rectifier potassium channel 1; delayed rectifier potassium channel Kv2.1; DRK1; DRK1PC; KCB1; Kcnb1; Kcr1-1; Kv2.1; mShab; potassium channel Kv2.1; potassium channel, voltage gated Shab-related subfamily B, member 1; potassium voltage gated channel, Shab-related subfamily, member 1; potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Shab; voltage-gated potassium channel subunit Kv2.1 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 16500, 25736, 3745 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.08% sodium azide | |
| P15387, Q03717, Q14721 | |
| Kcnb1 | |
| Synthetic peptide, conjugated to KLH, corresponding to the rat Kv2.1 potassium channel. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction