missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv12.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-14142
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Kv12.2 Polyclonal specifically detects Kv12.2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| Kv12.2 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| BEC1, brain-specific eag-like channel 1, ELK channel 2, ELK2, ether-a-go-go K(+) channel family member, ether-a-go-go-like potassium channel 2, KIAA1282, Kv12.2, potassium voltage-gated channel subfamily H member 3, potassium voltage-gated channel, subfamily H (eag-related), member 3, voltage-gated potassium channel subunit Kv12.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| KCNH3 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: EVDTSSLSGDNTLMSTLEEKETDGEQGPTVSPAPADEPSSPLLSPGCTSSSSAAKLLSPRRTAPRPRLGGRGRPGRAGALK | |
| 0.1 mL | |
| Signal Transduction | |
| 23416 | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto