missing translation for 'onlineSavingsMsg'
Learn More

Kv11.3 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18372385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372385 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372385 Supplier Novus Biologicals Supplier No. NBP306083AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kv11.3 Polyclonal specifically detects Kv11.3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv11.3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Eag-related protein 3, ERG-3, ERG3eag related protein 3, ether-a-go-go related gene potassium channel 3, Ether-a-go-go-related gene potassium channel 3, Ether-a-go-go-related protein 3, hERG-3, HERG3erg3, Kv11.3MGC45986, potassium channel subunit HERG-3, potassium voltage-gated channel subfamily H member 7, potassium voltage-gated channel, subfamily H (eag-related), member 7, Voltage-gated potassium channel subunit Kv11.3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Kv11.3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90134
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.